EasySep™ Direct Human Eosinophil Isolation Kit
Fournisseur: STEMCELL Technologies
19656
STMC19656EA 686 EUR InStock
STMC19656
EasySep™ Direct Human Eosinophil Isolation Kit
Cell Separation Media
Immunomagnetic isolation of eosinophils directly from whole blood
  • Effectively depletes RBCs without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 98% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human Eosinophil Isolation Kit isolates functional, highly purified eosinophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité