EasySep™ Mouse Epithelial Cell Enrichment Kit II
Fournisseur: STEMCELL Technologies
19868
STMC19868EA 1024 EUR InStock
STMC19868
EasySep™ Mouse Epithelial Cell Enrichment Kit II
Cell Separation Media
Immunomagnetic isolation of untouched mouse epithelial cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Mouse Epithelial Cell Enrichment Kit II is designed to isolate epithelial cells from freshly dissociated mouse mammary tissues by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted non-epithelial cells with biotinylated antibodies and magnetic particles. The magnetically labelled cells are separated from untouched desired cells by using an EasySep™ magnet and simply pouring the desired cells into a new tube.
This product replaces the EasySep™ Epithelial Cell Enrichment Kit (Catalog #19758) for even faster isolations. It also replaces the cell enrichment portion of the EasySep™ Mouse Mammary Stem Cell Enrichment Kit (Catalog #19757).
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité