DMEM with 4500 mg/L D-Glucose
Fournisseur: STEMCELL Technologies
36250
STMC36250EA 49 EUR InStock
STMC36250
DMEM with 4500 mg/L D-Glucose
Milieux de culture cellulaire
Dulbecco's Modified Eagle's Medium (DMEM) with 4500 mg/l D-glucose.
Dulbecco's Modified Eagle's Medium (DMEM) with 4500 mg/l D­glucose is recommended for a wide variety of cell culture applications. Selection of suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition required for the cell culture application.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité