Alpha MEM without Nucleosides
Fournisseur: STEMCELL Technologies
36453
STMC36453EA 59 EUR InStock
STMC36453
Alpha MEM without Nucleosides
Milieux de culture cellulaire
Minimum Essential Medium Eagle - alpha modification (Alpha MEM) without nucleosides.
Minimum Essential Medium Eagle - Alpha Modification (Alpha MEM) without Nucleosides is recommended for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition. MEM has typically been used for the cultivation of cells grown in monolayers; however, additional supplementation to Alpha MEM has expanded the applications for this medium.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité