Hanks' Balanced Salt Solution with 10 mM HEPES, without Phenol Red
Fournisseur: STEMCELL Technologies
37150
STMC37150EA 92 EUR InStock
STMC37150
Hanks' Balanced Salt Solution with 10 mM HEPES, without Phenol Red
Buffers Tampons de culture de tissus
Hanks’ Balanced Salt Solution (HBSS) can be used for diluting, washing, irrigating or as a transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions and a carbohydrate as an energy source. Selection of suitable nutrient medium is dependent on type of cell, culture conditions, and degree of chemical definition required for the cell culture application.
Species: Human, mouse, non human primate, other, rat
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité