Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5]

Fournisseur: STEMCELL Technologies
60017
STMC60017EA 265 EUR InStock
STMC60017
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5]
Anticorps
Rat monoclonal IgG2a antibody against mouse CD4, unconjugated.

  • T cells
  • T cells CD4
  • T cells Regulatory

The RM4-5 antibody reacts with mouse CD4, an ~55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T helper/inducer cells), and at lower levels on dendritic cells. In the mouse, CD4 is not expressed by monocytes/macrophages. CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex. CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck. Binding of the RM4-5 antibody to CD4 inhibits ligand binding in vitro; binding of the RM4-5 antibody can be blocked by the clone GK1.5 antibody. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852) and EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kit (Catalog #18782).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité