Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD49b Rat Monoclonal Antibody [clone: DX5] (Alexa Fluor® 488)

Fournisseur: STEMCELL Technologies
60020AD
STMC60020ADEA 136 EUR InStock
STMC60020AD
Anti-CD49b Rat Monoclonal Antibody [clone: DX5] (Alexa Fluor® 488)
Anticorps
Rat monoclonal IgM antibody against mouse CD49b (integrin α2), Alexa Fluor® 488-conjugated.

  • Nk cells

The DX5 antibody reacts with murine CD49b (integrin α2), an ~150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) to form the heterodimeric CD49b/CD29 complex known as VLA-2, a receptor for extracellular matrix proteins such as collagen, E-cadherin, fibronectin, and laminin. CD49b is highly expressed by platelets and is found on a majority of NK cells, on NKT cells, and on a small subset of CD8+ T cells; the latter population increases substantially following viral infection. CD49b is also expressed by several tissues, including intestine, kidney, mammary gland, and lung. The DX5 antibody is particularly useful for identifying NK cells in mice lacking the NK1.1 antigen. Binding of the DX5 antibody has not been observed to affect the function of the VLA-2 integrin. DX5 binding is, however, blocked by the clone HMα2 antibody. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD49b
Clonality: Monoclonal
Clone: DX5
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgM kappa
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité