Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD90 Mouse Monoclonal Antibody [clone: OX-7] (APC)

Fournisseur: STEMCELL Technologies
60024AZ
STMC60024AZEA 382 EUR InStock
STMC60024AZ
Anti-CD90 Mouse Monoclonal Antibody [clone: OX-7] (APC)
Anticorps
Mouse monoclonal IgG1 antibody against mouse, rat, guinea pig CD90/CD90.1, APC-conjugated.

  • T cells

The OX-7 antibody reacts with rat CD90 (Thy-1) and mouse The OX-7 antibody reacts with rat CD90 (Thy-1) and mouse CD90.1 (Thy-1.1), the latter being an allelic form of CD90 expressed by mouse strains AKR/J, PL, and FVB/N. The OX-7 antibody does not react with CD90.2, which is expressed by many mouse strains, including CBA and BALB/c. CD90 is a GPI-linked membrane glycoprotein and member of the immunoglobulin superfamily. The 25 kDa core protein is N-glycosylated at three sites, giving rise to molecules with a range of molecular masses (25 - 37 kDa). In the rat, CD90 is expressed by several cell types, including hematopoietic stem cells, immature B cells, thymocytes and neurons. In mouse strains expressing CD90.1, it is found on early-stage hematopoietic cells in the bone marrow, thymocytes, and circulating mature T cells. The OX-7 antibody has been reported to induce leukocyte activation, glomerular nephritis, apoptosis in glomerular mesangial cells, and vascular permeability. This antibody clone has been verified for purity assessments of cells isolated from compatible mouse strains with EasySep™ kits, including EasySep™ Mouse T Cell Isolation Kit (Catalog #19851).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD90
Clonality: Monoclonal
Clone: OX-7
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Guinea pig, Mouse, Rabbit, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité