Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8]

Fournisseur: STEMCELL Technologies
60031
STMC60031EA 314 EUR InStock
STMC60031
Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8]
Anticorps
Rat monoclonal IgG2a antibody against mouse Ly-6G, unconjugated.

  • Granulocytes
  • Subsets
  • Myeloid cells

The 1A8 antibody reacts with Ly-6G, a 21 - 25 kDa GPI-anchored protein, which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen (Gr-1). Gr-1 is expressed on monocytes, neutrophils and subsets of macrophages, plasmacytoid dendritic cells, and T cells. Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation. In the periphery, Gr-1 is found predominantly on neutrophils and is a useful marker for these cells. The 1A8 antibody binds specifically to Ly-6G, whereas another commonly used antibody, clone RB6-8C5, binds to both Ly-6G and Ly-6C. It has been reported that the 1A8 antibody detects Ly-6G-expressing granulocytes in peripheral blood, whereas the RB6-8C5 antibody also binds to Ly-6C-expressing lymphocytes, monocytes, and dendritic cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: Ly-6G
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité