Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD11b Mouse Monoclonal Antibody [clone: ICRF44] (Biotin)

Fournisseur: STEMCELL Technologies
60040BT
STMC60040BTEA 282 EUR InStock
STMC60040BT
Anti-CD11b Mouse Monoclonal Antibody [clone: ICRF44] (Biotin)
Anticorps
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD11b, biotin-conjugated.

  • Myeloid cells

The ICRF44 antibody reacts with an extracellular epitope on CD11b (integrin αM), an ~170 kDa type 1 transmembrane glycoprotein which associates non-covalently with CD18 to form the heterodimeric Mac-1 receptor. Through its interactions with ligands such as ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), iC3b, heparin, and fibrinogen, Mac-1 influences several processes, including the adherence of neutrophils and monocytes to stimulated endothelium, and phagocytosis of complement-coated particles. CD11b is expressed on the surface of granulocytes, monocytes, NK cells, dendritic cells, tissue macrophages, and subsets of T and B cells, and has been used as a marker to distinguish naïve and memory CD8+ T cells. CD11b is a relatively late marker for myeloid differentiation, and is undetectable on most myelomonocytic hematopoietic progenitor cells and more primitive cells. Certain mutations in CD11b give rise to the disorder systemic lupus erythematosus. The ICRF44 antibody reportedly inhibits leukocyte aggregation in response to the chemoattractant fMLP. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ human Monocyte enrichment kit (Catalog #19059), and for labeling human mesenchymal cells grown in MesenCult™ proliferation kit (human; Catalog #05411).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD11b Mouse Monoclonal Antibody [clone: ICRF44] (Biotin)
Antigen: CD11b
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Chimpanzee, Common marmoset, Cynomolgus monkey, Human, Pig, Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité