Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631]

Fournisseur: STEMCELL Technologies
60061.1
STMC60061.1EA 222 EUR InStock
STMC60061.1
Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631]
Anticorps
Rat monoclonal IgM antibody against human, mouse, rat SSEA-3, unconjugated.

  • Pluripotent stem cells

The MC-631 antibody reacts with a galactosylgloboside epitope on the stage-specific embryonic antigen-3 (SSEA-3), which is expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey ES cell lines. No immunoreactivity is evident with undifferentiated mouse EC, EG and ES cells. Expression of SSEA-3 is down regulated following differentiation of ES and EC cells. In contrast, the differentiation of mouse ES and EC cells may be accompanied by an increase in SSEA-3 expression. This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850) and TeSR™2 (Catalog #05860) and has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ human ES/iPS cell TRA-1-60 positive selection kit (Catalog #18166).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631]
Antigen: SSEA-3
Clonality: Monoclonal
Clone: MC-631
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM kappa
Reactivity: Human, Mouse, Rat, Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité