Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60R]

Fournisseur: STEMCELL Technologies
60064
STMC60064EA 357 EUR InStock
STMC60064
Anti-TRA-1-60 Mouse Monoclonal Antibody [clone: TRA-1-60R]
Anticorps
Mouse monoclonal IgM antibody against human, rhesus, rabbit TRA­1­60 (podocalyxin), unconjugated.

  • Pluripotent stem cells

The TRA‐1‐60R antibody reacts with TRA-1-60 (Podocalyxin), a >200 kDa pluripotent stem cell-specific protein expressed on the surface of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells, embryonal carcinoma (EC) cells and embryonic germ (EG) cells, as well as rhesus monkey ES cell lines. A soluble form of TRA-1-60 has been detected in serum of patients with embryonal carcinoma. The epitope, which is lost upon cell differentiation, contains sialic acid and is associated with a large-molecular-mass transmembrane protein named podocalyxin. Though sialylated, the epitope recognized by the TRA‐1‐60R antibody is resistant to treatment with neuraminidase. This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850), and TeSR™2 (Catalog #05860), and for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human ES/iPS Cell TRA-1-60 Positive Selection Kit (Catalog #18166; partial blocking may be observed) and EasySep™ hESC/hiPSC SSEA-4 Positive Selection Kit (Catalog #18165).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: TRA-1-60
Clonality: Monoclonal
Clone: TRA-1-60R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Rabbit, Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité