Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD40 Mouse Monoclonal Antibody [clone: 5C3] (Alexa Fluor® 488)

Fournisseur: STEMCELL Technologies
60113AD
STMC60113ADEA 369 EUR InStock
STMC60113AD
Anti-CD40 Mouse Monoclonal Antibody [clone: 5C3] (Alexa Fluor® 488)
Anticorps
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD40, Alexa Fluor® 488-conjugated.

  • B cells

The 5C3 antibody reacts with CD40, a 45 to 50 kDa type I transmembrane glycoprotein that is a member of the tumor necrosis factor receptor (TNFR) superfamily. CD40 plays an important role in B cell development and is also involved in the process of B cell selection in germinal centers. It facilitates T cell-B cell interactions for co-stimulation and immune regulation through engagement with CD154 (gp39), the CD40 ligand expressed on T cells. In the presence of IL-4, or other stimuli, CD40 promotes B cell proliferation, induces immunoglobulin- (Ig) isotype class switching and rescues B cells from surface Ig-induced apoptosis. CD40 is expressed on the surface of B cells, follicular dendritic cells, macrophages, fibroblasts, keratinocytes, endothelial cells, basal epithelial cells, CD34+ hematopoietic progenitor cells, and primary carcinomas. It is expressed at low levels, or not at all, on plasma cells. The 5C3 antibody has been used for the activation of B cells in in vitro functional assays or for the induction of B cell proliferation.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD40 Mouse Monoclonal Antibody [clone: 5C3] (Alexa Fluor® 488)
Antigen: CD40
Clonality: Monoclonal
Clone: 5C3
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Chimpanzee, Cynomolgus monkey, Human, Rhesus monkey, Squirrel monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité