Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD24 Mouse Monoclonal Antibody [clone: 32D12] (Biotin)

Fournisseur: STEMCELL Technologies
60134BT
STMC60134BTEA 854 EUR InStock
STMC60134BT
Anti-CD24 Mouse Monoclonal Antibody [clone: 32D12] (Biotin)
Anticorps
Mouse monoclonal IgG1 antibody against human CD24, biotin-conjugated.

  • B cells

The 32D12 antibody reacts with CD24, a 35 to 50 kDa glycosylphosphatidylinositol (GPI)-linked sialoglycoprotein that is anchored to the plasma membrane and is glycosylated variably depending on cell type. The function of CD24 has yet to be fully determined but it is believed to play a role in lymphocyte development, particularly in acting as a costimulatory molecule for CD4+ T cells and in regulating B cell proliferation, activation, differentiation, and apoptosis. CD24 may also function in cell adhesion and metastasis, likely through binding of CD62P (P-Selectin). CD24 is expressed by follicular dendritic cells, and various epithelial and hematopoietic cell types, including granulocytes, erythrocytes, most B cells, and subsets of T lymphocytes. As it is expressed throughout B cell development but is lost after differentiation into the plasma cell stage, CD24 may be used as a B cell differentiation marker. Furthermore, CD24 also has the potential to be a marker for tumor prognosis and diagnosis as it is highly expressed in numerous cancers including ovarian, breast, prostate, bladder, renal, and lung, as well as neuroblastomas and various hematological malignancies. Reportedly, the 32D12 antibody recognizes an epitope that is distinct from those recognized by clones VIB-C5, VIB-E3, LC66, BA-1, ML5, OKB2 and SN3 A5-2H10.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD24 Mouse Monoclonal Antibody [clone: 32D12] (Biotin)
Antigen: CD24
Clonality: Monoclonal
Clone: 32D12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité