Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD326 (EpCAM) Mouse Monoclonal Antibody [clone: VU-1D9] (FITC)

Fournisseur: STEMCELL Technologies
60136FI
STMC60136FIEA 231 EUR InStock
STMC60136FI
Anti-CD326 (EpCAM) Mouse Monoclonal Antibody [clone: VU-1D9] (FITC)
Anticorps
Mouse monoclonal IgG1 antibody against human CD326 (EpCAM), FITC-conjugated.

  • Mammary cells

The VU-1D9 antibody reacts with the human epithelial cell adhesion molecule (EpCAM or CD326), an ~40 kDa type I transmembrane glycoprotein, which functions as a homophilic and Ca++-independent adhesion molecule. Binding of the VU-1D9 antibody requires the presence of the extracellular EGF-1 domain of EpCAM. EpCAM has roles in several cellular processes, including signaling, migration, proliferation, and differentiation. It is expressed early during erythrogenesis and has been identified as a stem cell marker. The EpCAM protein is found on the basolateral membrane of most normal epithelial cells and is highly expressed in bone marrow, colon, and lung. EpCAM is not expressed on mesothelial cells or mesotheliomas, and so is widely used as a diagnostic marker to distinguish mesothelioma and carcinoma cells, as well as serving as a target for immunotherapeutic treatment of carcinomas. The VU-1D9 antibody has been used to identify and isolate circulating tumor cells.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD326 (EpCAM)
Clonality: Monoclonal
Clone: VU-1D9
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité