Human Recombinant IL-22
Fournisseur: STEMCELL Technologies
78038
STMC78038EA 538 EUR InStock
STMC78038
Human Recombinant IL-22
Protéines et Peptides
Interleukin 22 (IL-22) is a class 2 α-helical cytokine that signals through the class 2 cytokine receptor and activates the JAK/STAT pathway. IL-22 is secreted by Th1, Th2, Tc22, and γδ T cells, dendritic, mast and NK cells. It stimulates expression of antimicrobial peptides from epithelial cells, thus hindering bacterial infections. Depending on the interactions with other cytokines, IL-22 can either promote inflammation or prevent tissue destruction by regulating host defense and tissue homeostasis at barrier surfaces (Sonnenberg et al.).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité