Mouse Recombinant TRAIL
Fournisseur: STEMCELL Technologies
78122.1
STMC78122.1EA 178 EUR InStock
STMC78122.1
Mouse Recombinant TRAIL
Protéines et Peptides
TRAIL (TNF-related apoptosis-inducing ligand) belongs to the tumor necrosis factor (TNF) superfamily and is associated with initiating apoptosis. TRAIL has four major receptors: two death receptors DR4 and DR5, and two decoy receptors DcR1 and DcR2. TRAIL binds to the death receptors, which recruits the Fas-associated death domain (FADD) and activates caspases 8 and 10, which eventually leads to apoptosis (Pitti et al.; Wiley et al.; Zauli and Secchiero). It has been shown that mice lacking the expression of TRAIL have defects in thymocyte apoptosis and negative selection, and these mice had increased susceptibility to autoimmune diseases (Lamhamedi-Cherradi et al.).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité