Human Recombinant EGFR
Fournisseur: STEMCELL Technologies
78171.1
STMC78171.1EA 373 EUR InStock
STMC78171.1
Human Recombinant EGFR
Protéines et Peptides
Epidermal growth factor receptor (EGFR) is a type I transmembrane protein and receptor tyrosine kinase. EGFR has been shown to bind to some members of the EGF family ligands including EGF, amphiregulin, TGF-α, betacellulin, epiregulin, heparin-binding EGF, and neuregulin-2α. EGFR ligand binding induces homodimerization, as well as heterodimerization of EGFR with ErbB2 or with ligand-activated ErbB3 or ErbB4 (Schlessinger). Dimerization results in kinase activation, phosphorylation, and cell signaling, mediated primarily through MEK/ERF and AKT pathways (Navlonic et al.). EGFR signaling has been shown to regulate cell proliferation, differentiation, motility, and apoptosis. Elevated levels of EGFR have been correlated with carcinogenesis (Maihle et al.). Protein contains a His-residue tag at the carboxyl end of the polypeptide chain.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité