Human Recombinant FGF-5
Fournisseur: STEMCELL Technologies
78182.1
STMC78182.1EA 273 EUR InStock
STMC78182.1
Human Recombinant FGF-5
Protéines et Peptides
Fibroblast growth factor 5 (FGF-5) is a secreted, heparin-binding member of the FGF subfamily. FGFs possess broad mitogenic and cell survival activities and are expressed during embryonic development. FGF-5 is expressed in the mesenchyme, skeletal muscles, central nervous systems, and hair follicles. FGF-5 promotes cell differentiation and proliferation by binding FGF receptor 1 and FGF receptor 2 (FGFR1 and FGFR2, respectively). FGF-5 plays an important regulatory role in skeletal muscle development. FGF-5 has also been identified in neurons of the limbic system, especially those of the olfactory bulb and pyramidal cells of the hippocampus (Haub and Goldfarb).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité