Human Recombinant FGF-6
Fournisseur: STEMCELL Technologies
78184.1
STMC78184.1EA 1593 EUR InStock
STMC78184.1
Human Recombinant FGF-6
Protéines et Peptides
Fibroblast growth factor 6 (FGF-6) is a heparin-binding member of the FGF family, regulators of cell proliferation, differentiation, and function. FGF-6 binds and signals through the FGF receptors 1c, 2c, and 4 (Ornitz et al.). FGF-6 is a potent mitogen for fibroblasts, vascular endothelial cells, and prostate carcinoma cells (Asada et al.; Pizette et al.; Ropiquet et al.). FGF-6 is primarily expressed in epithelial and mesenchymal cell lineages. During development, FGF-6 is expressed in skeletal muscle, consistent with its role in muscle differentiation and regeneration (Floss et al.). FGF-6 has also been shown to promote chondrogenesis in embryonic somites in conjunction with transforming growth factor beta 2 (TGF-β2; Grass et al.).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité