Synonyms:
Hydrogen oxide
79001
79002
79002.1
STMC79001EA
59
EUR
InStock
STMC79001
STMC79002
STMC79002.1
Eau sans nucléase
Eau
DNase- and RNase-free water for molecular biology applications.
Nuclease-Free Water is used in various molecular biology applications requiring nuclease-free conditions, such as in processing DNA or RNA (i.e. PCR, cDNA synthesis, or qPCR). This product is sterile-filtered (0.2 µm), free of RNase and DNase activity, endotoxin-free, and not DEPC-treated.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').