Gel and PCR Clean-up Kit
Fournisseur: STEMCELL Technologies
79030
STMC79030EA 676 EUR InStock
STMC79030
Gel and PCR Clean-up Kit
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
For extraction of DNA from agarose gels, or clean-up of PCR products and restriction enzyme digestions.
The Gel and PCR Clean-up Kit uses a membrane-based system to quickly purify DNA fragments of 100 bp to 10 kb in length from polymerase chain reaction (PCR), restriction enzyme digestion, or excised agarose gel fragments (up to 350 mg). The purified DNA is directly suitable for applications such as automated fluorescent DNA sequencing, cloning, labelling, restriction enzyme digestion, or in vitro transcription. Up to 40 µg of DNA can be processed per column in as little as 20 minutes and eluted in volumes as low as 15 µl, with up to 95% recovery depending on DNA fragment length. The Gel and PCR Clean-up Kit can be used to purify linear DNA fragments, supercoiled plasmid DNA, or single-stranded DNA (circular or linear).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité