MethoCult™ Cell Wash Medium
Fournisseur: STEMCELL Technologies
87700
STMC87700EA 70 EUR InStock
STMC87700
MethoCult™ Cell Wash Medium
Milieux de culture cellulaire
Medium for preparing and washing samples for colony-forming unit (CFU) assays.
MethoCult™ cell wash medium is recommended for the preparation and washing of human bone marrow, peripheral blood or cord blood samples prior to performing colony-forming unit (CFU) assays using MethoCult™ medium.
Please note: In light of the new EU regulation 2017/746 (IVDR) of the european parliament and of the council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled 'for research use only'. Please note there have been no changes to the form, function and quality assurances associated with these products.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité