Cole-Parmer® HP-200 Series Large-Capacity Digital Hotplate
Fournisseur: Cole Parmer
04807-82
STUA04807-82EA 1620 EUR InStock
STUA04807-82
Cole-Parmer® HP-200 Series Large-Capacity Digital Hotplate
Appareils chauffants Plaques chauffantes
Microprocessor controlled, dual thermocouples ensure accurate temperature control.
  • Push buttons and dial setting for fast temperature changes and simple control
  • Large plate area accommodates one large vessel or several smaller ones
  • Be safe - hot warning light remains active even when unplugged while above 50 °C
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
Microprocessor control allows for accurate plate temperature monitoring. The ‘Hot’ warning light will flash whenever the plate temperature is above 50 °C. AnMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité