Temperature controller, accessory for hotplates and stirrers, Stuart® SCT2, Cole-Parmer
Fournisseur: Cole Parmer
04807-90
STUA04807-90EA 421 EUR InStock
STUA04807-90
Temperature controller, accessory for hotplates and stirrers, Stuart® SCT2, Cole-Parmer
Contrôleurs Contrôleurs de température
All hotplate models are compatible with the Stuart temperature controller SCT2. The SCT2 provides precise temperature control in aqueous or oily samples up to a maximum temperature of 200 °C.
  • Automatically recognized when connected
  • Accurate setting of liquid temperature
  • Built-in holder for support rod and probe holder
  • Removable temperature sensor
  • Bright, easy-to-read LED display
  • Compact and lightweight
Designed for use with STUART UC and US range magnetic hotplate stirrers.
A range of accessories is available to complete your equipment: Support rod; PTFE probe (for applications requiring chemical resistance) and a metal probe.
Information de livraison: Supplied with stainless steel temperature probe.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité