Cole-Parmer® WB-300 Series Shaking Water Baths, Stuart
Fournisseur: Cole Parmer
12152-02
STUA12152-02EA 3670 EUR InStock
STUA12152-02
Cole-Parmer® WB-300 Series Shaking Water Baths, Stuart
Bains Bains-marie à agitation
Customisethe shaking water bath to your requirements with platforms and racks
  • Analogue controls and digital temperature display simplify operation
  • Over temperature and low level cut-offs improve safety
  • Shaking speed: 20 to 130 min⁻¹
  • Stroke length/orbit size: ³/₄" (20 mm)
This water bath can be used as a linear or orbital shaker, or as a static bath by simply changing the platform. Strong magnets couple to the drive unit so platform changes are quick and easy. The stainless steel shaking platforms with high-tension springs accommodate most bottle and flask sizes. It can hold up to 8×250, 6×500 ml, or 4×1 L Erlenmeyer flasks. For tubes and smaller vessels, order an optional stainless steel rack.
Set speed and temperature using analogue dials. Use the LED display to set and view current temperature. A polycarbonate cover (12152-99) or stainless steel cover is required to reach temperatures above 90 °C (order separately). Stainless steel tank with integral drain makes cleaning and maintenance fast and efficient.
Information de livraison: Supplied with power cords with UK and Shuko plug.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité