Cole-Parmer® Essentials Bottle Top Dispensers with Recirculation Valve
Fournisseur: Cole Parmer
25208-31
STUA25208-31EA 763 EUR InStock
STUA25208-31
Cole-Parmer® Essentials Bottle Top Dispensers with Recirculation Valve
Distributeurs pour flacons
Save money by conserving reagents.
  • Recirculation valve prevents loss of reagent during priming and purging
  • Smooth plunger movement provides bubble-free dispensing
  • Adjustable nozzle allows users to change the dispensing angle
  • Wetted materials: PTFE, ETP, borosilicate glass, PP, PFA, FEP
  • Autoclavable
These bottle top dispensers conserve reagents with a built-in recirculation valve, allowing users to prime and purge without waste. The PTFE piston with ETP O-ring provide frictionless movement, reducing the occurrence of bubbles in dispenses. The springless PTFE valve manifold offers superior chemical compatibility and resists jamming.
Information de livraison: Includes discharge tube, telescoping filling tube, calibration tool, bottle adapter set (28, 32, 38, 40, and 45 mm) and calibration certificate.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité