Extractables and Leachables Screening Standard for GC, certified reference material, TraceCERT®, 14 components in tert-Butyl methyl ether, Supelco®
Fournisseur: Merck
01829-1ML
SUPL01829-1MLEA 266 EUR InStock
SUPL01829-1ML
Extractables and Leachables Screening Standard for GC, certified reference material, TraceCERT®, 14 components in tert-Butyl methyl ether, Supelco®
Étalons Étalons organiques
Ready to use certified reference material mixture suitable for dentification and quantification of extractables and Leachables by GC. 14 components in a concentration of 50 μg/mL.
  • Certification of each individual component by qNMR (following ISO 17025 accreditation)
  • Mixes produced following the ISO 17034 workflow
  • Tested for homogeneity and long-term stability using GC-MS
  • Traceability to NIST SRM
  • Supplied with a comprehensive certificate including the overall uncertainty
Components:
Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate
ε-Caprolactam
2-Mercaptobenzothiazole
Bisphenol A
3,5-Di-tert-4-butylhydroxytoluene (BHT)
1,3-Di-tert-butylbenzene
Oleamide
Bis(2-ethylhexyl) phthalate
Stearic acid
cis-13-Docosenoamide
Tris(2,4-di-tert-butylphenyl)phosphate
2,4-Di-tert-butylphenol
2,6-Di-tert-butylphenol
Palmitic acid
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité