Scalpel handle, Cygnetic®
Fournisseur: SWANN-MORTON
5398
SWAN5398EA 143 EUR InStock
SWAN5398
Scalpel handle, Cygnetic®
Bistouris et pièces détachées Manches de bistouri
This stainless steel handle is designed to assist the OR nurse with safe and easy blade attachment and removal before, during and after the procedure.
  • Safe and easy blade attachment and removal
  • Simple disassembly for cleaning
  • Great strength and stability
  • Handle is non sterile, blades are sterile
Ideal for use within rigorous Orthopaedic and general surgical procedures.
Conditionnement: Handle is supplied individually sleeved. Blades (to be ordered separately) are supplied individually V.P.I. protected in metal foil packets.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité