Colony counter, aCOLade 2
Fournisseur: SYNBIOSIS
ACOLAD2
SYNBACOLAD2EA 1430 EUR InStock
SYNBACOLAD2
Colony counter, aCOLade 2
Compteurs de colonies
aCOLade2 is designed with an antimicrobial protection. The pressure of marking a colony with a felt-tip pen registers a cumulative count. The pressure can be adjusted to each user.
  • Audible count confirmation (can be turned on or off)
  • Choice of light or dark background, illuminated receiver plate
  • Switchable black background to enhance translucent and difficult to see colonies
Sub-stage illumination by low energy bright LEDs allows glare-free optimum viewing. A switchable black background is provided to enhance viewing of translucent and difficult to see colonies. Supplied with two graticules and dish centering adapters to facilitate use with 50 - 90 mm dishes.
Informations de commande: A choice of magnifiers and a printer are available as optional accessories.
Information de livraison: Supplied with two graticules and dish centering adapters for Petri dishes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité