ENERGY Easytabs for Dishwashers
Fournisseur: WERNER & MERTZ BENELUX SA NV
715844
TANA715844EA 218 EUR InStock
TANA715844
ENERGY Easytabs for Dishwashers
Produits nettoyants Détergents et Nettoyants de Surfaces
ENERGY easytabs four in one tabs for dishwashers.
  • Multi-performing
  • Easy dosage
  • Material friendly
Detergent: Removes stains and grease and makes your dishes clean
Rinse aid: Accelerates drying time, prevents lime scale deposits, and makes dishes sparkle
Regeneration: Prevents the formation of white veil, for bright dishes
Protection: Protects your dishes, for a longer lifespan
Meanwhile enhancing the work safety, the product is phosphate free and fragrance free and prevents scaling of the heating elements.
Majorly composed of renewable resources, ENERGY easytabs takes on the responsibility for future generations.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité