A3218-250ML
TCIAA3218-250MLEA
45.1
EUR
InStock
TCIAA3218-250ML
Acrylamide : Bisacrylamide (37.5:1) 30% dans l'eau pour l'électrophorèse
Acrylamide : Bisacrylamide
30% (w/w) Acrylamide / Bis-acrylamide solutions are available at the most common ratios for use in protein electrophoresis.
Convenient to use, reduced risk of neurotoxic acrylamide dust in the air.