Synonyms:
Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate
A5161-5G
TCIAA5161-5GEA
20.9
EUR
InStock
TCIAA5161-5G
1-Cétylpyridinium chlorure monohydraté ≥98.0% (par HPLC, analyse par titration)
Chlorure de 1-cétylpyridinium monohydraté
Ion association reagent for extraction photometric analysis and sensitizer in photometric determination of metals.