Synonyms:
Dimethylaniline
D0664-25ML
TCIAD0664-25MLEA
18.7
EUR
InStock
TCIAD0664-25ML
Xylidine mélange >65.0% (par GC)
Xylidine
Molecular Weight: 121.18
Purity/Analysis Method: 65.0% (GC)
Form: Clear Liquid
Boiling point (°C): 213
Flash Point (°C): 97
Specific Gravity (20/20): 0.98