Synonyms:
Dibutyldisulphide, n-Butyl disulphide
D1395-25ML
TCIAD1395-25MLEA
31.9
EUR
InStock
TCIAD1395-25ML
Disulfure de dibutyle ≥98.5% (par GC)
Disulfure de dibutyle
Reagent for Determination of Sulfur Content by Energy Dispersive X-ray Fluorescence Method.