Synonyms:
N-Fluoro-N-(tert-butyl)-tert-butanesulphonamide, N-tert-Butyl-N-fluoro-2-methyl-2-propanesulphonamide, NFBB
F1345-1G
TCIAF1345-1GEA
176
EUR
InStock
TCIAF1345-1G
N-(tert-Butyl)-N-fluoro-2-methylpropane-2-sulphonamide ≥95.0% (par GC), Solide
N-(tert-Butyl)-N-fluoro-2-methylpropane-2-sulphonamide
Easy-to-handle and Mild Electrophilic Fluorinating Agent NFBB
- Enables fluorination of active methylene compounds, (hetero)aryl lithium and alkenyl lithium species, Grignard reagents
- Achieves high yield fluorination due to the reduction of side reactions by tert-butyl groups
- Higher fluorine content than conventional fluorinating agents
- Applicable to the synthesis of pharmaceutical intermediates