Synonyms:
8-Methyl-N-vanillyl-6-none-amid
M1149-1G
TCIAM1149-1GEA
64.9
EUR
InStock
TCIAM1149-1G
Capsaicine >60.0% (par HPLC)
Capsaicine
CAS Number: 404-86-4
MDL Number: MFCD00017259
Molecular Formula: C18H27NO3
Molecular Weight: 305.42
Purity/Analysis Method: 60.0% (HPLC)
Form: Crystal
Melting point (°C): 65
Flash Point (°C): 113
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Reagents for Pain & Itch Models, Reagents for Inflammation & Immunity Disorder Models, Alkaloids (Phytochemicals), Alkaloids (Others)
Other Product Names / Synonyms: N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methyl-6-nonenamide, 8-Methyl-N-vanillyl-6-nonenamide