Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
P0495-25ML
TCIAP0495-25MLEA
148
EUR
InStock
TCIAP0495-25ML
β-Methylstyrene (cis- and trans- mixture) mélange cis- et trans ≥95.0% (par GC) stabilisé
beta-Methylstyrene (cis- and trans- mixture)