Synonyms:
Hexamethyldisilazane and Trimethylchlorosilane in Anhydrous Pyridine, Trimethylchlorosilane and Hexamethyldisilazane in Anhydrous Pyridine
T0274-12ML
TCIAT0274-12MLEA
35.2
EUR
InStock
TCIAT0274-12ML
TMS-HT (=HMDS and TMCS in anhydrous pyridine) Trimethylsilylating Reagent, for OH compounds
TMS-HT (=HMDS and TMCS in anhydrous pyridine)