Tall Stainless Steel Storage Cupboards
Fournisseur: TEKNOMEK
SHCCTCSDDS1040
TEKMSHCCTCSDDS1040EA 0 EUR InStock
TEKMSHCCTCSDDS1040
Tall Stainless Steel Storage Cupboards
Mobilier Armoires de stockage
Stainless steel cupboards 304-grade, with a range of door options ensure robustness, hygiene and ease of cleaning. Designed with hygeine first!
  • Sloping top
  • Shelves sold separately
  • 6 to 8 easy clean shelve adjustments (every 180 mm)
  • Brushed dull polish finish
  • Stainless steel adjustable feet with non-marking rubber base
Supplied with 6 (1780 mm high) easy clean shelf adjustments. The shelves have been cleverly designed so any liquid will fall to the back of the cupboard and out through the drainage holes placed in the base of the unit.
The doors are extremely tough, manufactured from 1,5 mm thick stainless steel, set in a super strong frame making these cupboards incredibly robust.
The cupboards are designed with hygiene in mind too! No ledges or crevices assures that dirt cannot collect.
The solid stainless steel construction and sloping top enable easy cleaning and the 300 mm legs allow easy access for cleaning underneath.
Information de livraison: Supplied without shelves.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité