Infrared thermometers, testo 835-2
Fournisseur: TESTO
0560.8352
TEST0560.8352EA 497 EUR InStock
TEST0560.8352
Infrared thermometers, testo 835-2
Thermomètres Thermomètres numériques
Ideal for use in the glass, ceramics or the metal industry: the testo 835-T2 high temperature infrared thermometer is the ideal professional tool for a wide variety of thermal applications in the high temperature range. The testo 835-T2 has an extremely wide temperature range which allows the user to carry out thermal measurements up to 1 500 °C - and all from a safe distance. It doesn’t matter whether the objects are small, hard to reach, moving or very hot.
  • Extended temperature range for reliable measurements up to 1 500 °C
  • IR measurements from a safe distance; precise measuring of moving objects
  • Memory for up to 200 readings; easy analysis with free-to-download software
  • The 4-point laser clearly indicates the measuring area and prevents incorrect measurements
  • The 50:1 optics allow the user to carry out measurements from a long distance and still obtain excellent results (distance 5 m = 10 cm measuring spot)
  • The backlit display allows to use the infrared thermometer in all lighting conditions
For easy analysis and easy use: The testo 835-T2 comes with an easy-to-follow menu, self-explanatory icons and a user-friendly joystick. It can be used to create different measuring sites or save up to 200 readings while additional features including min/max values, adjustable thresholds and visual and acoustic alarms which are triggered when thresholds have been exceeded make work more efficient and life easier. The free-to-download software provides you with a quick and convenient means of analyzing the measuring data.
Certifications: EN 61326-1:2006
Information de livraison: testo 835-T2 high temperature infrared thermometer, 4-point laser marking, measuring data administration, batteries, calibration certificate.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité