Differential pressure manometer, testo 510
Fournisseur: TESTO
0563.0510
TEST0563.0510EA 165 EUR InStock
TEST0563.0510
Differential pressure manometer, testo 510
Manomètres
The pocket-sized testo 510 measures differential pressure in the range from 0 to 100 hPa. The differential pressure measurement is temperature-compensated for accurate measurement values.
  • Handy, small and easy to operate
  • 10 selectable units
  • Values can be displayed in Pascal over the entire measurement range
  • Temperature and air density compensation
  • Magnets at the rear permit free-hand work
  • Backlit display allows the measurement values to be easily read out, even in unfavourable light conditions
  • Flow velocity measurement possible with Pitot tube
Selectable units: hPa, mbar, Pa, mmH₂O, inH₂O, inHg, mmHg, psi, m/s, fpm
Power supply: Two AAA micro batteries (50 h battery life (average, without display illumination))
Information de livraison: Supplied with protection cap, batteries, belt holder, calibration protocol, and hoses.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité