delta P air flow ComboKit 2 with Bluetooth®, Testo 440
Fournisseur: TESTO
0563.4410
TEST0563.4410EA 1649 EUR InStock
TEST0563.4410
delta P air flow ComboKit 2 with Bluetooth®, Testo 440
Enregistreurs de données
Delta P air flow ComboKit 2 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
  • Carry out measurements without any annoying cables
  • With space-saving and integrated differential pressure sensor
  • Easily transported
Convenient with wireless probes, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality and maximum flexibility for duct and outlet measurements.
Information de livraison: Supplied with testo 440 dP air velocity and IAQ measuring instrument including differential pressure sensor, two vane probe heads (Ø 100 mm and Ø 16 mm) and a humidity/temperature probe head – all three can be attached to the universal Bluetooth® handle.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité