LED stroboscope, testo 477
Fournisseur: TESTO
0563.4770
TEST0563.4770EA 930 EUR InStock
TEST0563.4770
LED stroboscope, testo 477
Tachymètres
The hand-held LED stroboscope measures rotations and vibrations and facilitates measurements during operation. The stationary image enables the inspection and quality assessment of high-frequency moving parts.
  • Measurement range: Up to 300000 flashes per minute (fpm)
  • With light intensity of up to 1500 Lux
  • Trigger input and output enable connection to external systems and control by an external sensor
  • Long operating times: battery power for up to 5 h
The testo 477 LED stroboscope can also measure the rotational speed of very small and hard-to-reach objects. The integrated impact protection and sturdy IP 65 rated housing (which also allows it to be wiped down) ensure that the testo 477 can stand up to the rough and tumble of everyday use.
Dimensions: 191×82×60 mm
Information de livraison: Supplied with trigger signal plug, calibration protocol, batteries and carry case.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité