Infrared thermometer, 830
Fournisseur: TESTO
0560.8316 0563.8315
TEST0560.8316EA 153 EUR InStock
TEST0560.8316 TEST0563.8315
Infrared thermometer, 830
Thermomètres Thermomètres numériques
The testo 831 is so fast that measurements on palettes or refrigerated shelves can be carried out in a few seconds. Small objects such as yoghurt pots can be measured at a distance. A two-point laser indicates the exact measurement spot.
  • Adjustable emissivity of 0,2 to 1,0
  • 30:1 optics (measurement spot Ø is 36 mm at a distance of one metre)
  • Alarm limit values can be set and are optically and audibly indicated
Certifications: HACCP conform
Information de livraison: Supplied with a belt holder, a nine-volt battery for 15 hours of use, and a factory calibration certificate.
The set includes the testo 831 infrared thermometer and the testo 106 penetration thermometer, incl. TopSafe, belt holder and batteries.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité