Digital multimeter, testo 760-3
Fournisseur: TESTO
0590.7603
TEST0590.7603EA 218 EUR InStock
TEST0590.7603
Digital multimeter, testo 760-3
Compteurs électriques Multimètres numériques
The testo 760-3 digital multimeter is the multitasker for all professionals in the electricity sector. It enables users to carry out both simple and demanding electrical measuring tasks more reliably than ever before. The testo 760-3 automatically detects the appropriate measurement parameter via the socket assignment and therefore prevents dangerous incorrect settings.
  • Easy, reliable work thanks to automatic measurement parameter detection
  • Auto power off saves battery life
  • Rugged meter with overmould housing
  • Comfortable, one-handed operation without any dial
  • Large, backlit, dual readout display with analog bar graph is easy to read
  • Lead holder conveniently moulded into housing for easy storage
A voltage range of up to 1000 V, a frequency range of up to 30 MHz and capacitance of up to 60000 µF mean the testo 760-3 multimeter can also be used for industrial applications, such as troubleshooting in large electrical systems. And the integrated low-pass filter achieves more accurate measurement results on frequency converter controlled motors.
Certifications: EN 61326-1
Information de livraison: Supplied with test leads, three AAA batteries, instruction manual, quick start guide, and certificate of conformity.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité