Cleanroom bond paper, TexWrite® MP 10
Fournisseur: Texwipe
TX5800
TEXWTX5800EA 862 EUR InStock
TEXWTX5800
Cleanroom bond paper, TexWrite® MP 10
Papiers
TexWrite® MP 10 is a 100% synthetic substrate provides extremely low levels of particle generation along with superior usability. It is designed to use with ultraclean manufacturing environments (paperless) that cannot tolerate cellulose contamination.
  • Excellent ink capture with no risk of smearing
  • Low ESD potential for reduced risk of electrostatic damage
  • Compatible with inkjet, laser and toner-based printers and photocopiers
  • Ballpoint pens glide easily over surface, without skips or jumps
  • Autoclavable and recyclable as a other types of plastic
  • Distinctive blue coating on one side
Conditionnement: 100 sheets per ream and 5 ream per box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité