Automated stainers, Cellstain® 6
Fournisseur: THARMAC GmbH
JC651-30
THARJC651-30EA 10500 EUR InStock
THARJC651-30
Automated stainers, Cellstain® 6
Coloration pour lames
Automated stainer with six staining troughs for all staining protocols with up to six different chemicals (May-Grünwald, Giemsa, Wright, Pappenheim, Leishman, Gram staining).
  • Pre-programmed with six staining programs
  • Programs protected by a password
  • Very low consumption of stains and reagents
  • The agitation and the drain time is freely selectable
  • Linear staining machine to move the slide holder forward and backward
Suitable for haematology and bacteriology staining. Due to its compact size, it can be placed under a fume hood; a compatible fume extraction desk hood for vacuuming less hazardous pollutants during staining is available.
Informations de commande: Supplied with six staining troughs with lid, slide holder and shortened trough for loading and unloading.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité