Savant™ SpeedVac™ SPD2030A Integrated Vacuum Concentrator System
Fournisseur: THERMO SAVANT
SPD2030A-230
THSASPD2030A-230EA 28300 EUR InStock
THSASPD2030A-230
Savant™ SpeedVac™ SPD2030A Integrated Vacuum Concentrator System
Concentrateurs
Thermo Scientific™ Savant™ SpeedVac™ SPD2030 vacuum concentrators are fully integrated systems that combine a concentrator, refrigerated cold trap, diaphragm pump and a vacuum gauge into a single, compact unit.
  • Proven reliability: Over 50 years of experience with an oil-free PTFE diaphragm vacuum pump for virtually maintenance-free operation
  • Speed and efficiency: Rapid evapouration and solvent removal with a radiant cover and vacuum controls to prevent sample bumping
  • Ease of use: Intuitive design with a bright display, three preset and three user-defined
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité