VACUUM CLEANER MODEL EMI-CWR 120V - INCLUDES ACCESSORIES(S/S WANDS & HOSE ENDS) Technical features: Volt: 120V Hertz: 60 Hz Watt: 1 Kw Ampere: 8.5A Dry recovery Air Flow: 204m /h Vacuum at sealed orifice: 2413 mmH2O Recovery capacity: Dry: with 1 * 1 Pce
Fournisseur: TIGER-VAC
VACUUM CLEANER MODEL EMI-CWR 120V - INCLUDES ACCESSORIES(S/S WANDS & HOSE ENDS) Technical features: Volt: 120V Hertz: 60 Hz Watt: 1 Kw Ampere: 8.5A Dry recovery Air Flow: 204m /h Vacuum at sealed orifice: 2413 mmH2O Recovery capacity: Dry: with 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
VACUUM CLEANER MODEL EMI-CWR 120V - INCLUDES ACCESSORIES(S/S WANDS & HOSE ENDS) Technical features: Volt: 120V Hertz: 60 Hz Watt: 1 Kw Ampere: 8.5A Dry recovery Air Flow: 204m /h Vacuum at sealed orifice: 2413 mmH2O Recovery capacity: Dry: with 1 * 1 Pce TIGV110110A 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou